PrepTube for Sample Preparation, Distek, Inc.
: Avantor
For use with Ditek's PrepEngine. The PrepTube is polypropylene with SS blades and includes a cap with septa.
- 50, 250 or 500 ml volumes
- Clear or amber
- 10 preptubes per pack
The recommended usage for a single PrepTube is a maximum of 5×10 min tests or equivalent. Note: All tests are application specific.
Frequently Bought Together