T-Mop™ Adapters, Micronova
Supplier: Micronova
Certificates
About this item
Electropolished stainless steel frame with Delrin® plastic fittings, quick disconnect adapter, and 180° swivel action.
- Electropolished stainless steel
- For use with the T-Mop™ series
- Attach to a Universal handle
- Autoclavable
- Good compatibility with strong disnfectants and common detergents
Designed for use with T-Mop™ series covers. Attach to a universal handle. Designed for cleanroom use.
Frequently Bought Together