Model 2500 Select Bathless Dissolution System, Distek
Supplier: Avantor
Certificates
About this item
The model 2500 select bathless dissolution system offers patented bathless technology with short media heating times and smaller energy use. It can be configured as USP Apparatus 1, 2, 5 and 6 plus intrinsic dissolution. The extensive users, method and reporting storage capabilities are enabled by a brilliant touch screen.
- Bathless heating
- Wireless in-shaft temperature sensors
- Serialized component tracking
- Color touch screen display
- Electronic qualification guidance
- Quick change paddles and baskets
- USP compliant
Patented wireless temperature sensors continuously monitor and display the in-vessel temperature for each vessel. Unparalleled control of the instrument plus automatic storage of component serial numbers and automatic qualification reminders make the Model 2500 Select robust and reliable enough for QC applications yet flexible enough for R&D use.
Certifications: ETL, CE.
Ordering information: Does not include installation or qualification services.
Delivery: Includes shafts, vessels, paddles, baskets, basket adapters, vessel covers, and a one-year comprehensive warranty.