Bond Elut CN (Cyano) is a medium-polarity sorbent with many uses, particularly for compounds that would be irreversibly retained on C18 and C8 sorbents.
- Ideal for extracting aqueous analytes
- Retention in aqueous and organic matrices
- Useful for many applications
A medium polarity sorbent with many uses, Bond Elut Cyano (CN) is ideal for applications in which extremely nonpolar compounds would be irreversibly retained on high carbon load sorbents, such as C8 and C18. This endcapped version of the cyano sorbent is best utilized when extracting analytes from an aqueous matrix.