Orders LinkContactUsLinkComponent
Omnimed® Mounting Magnets
Mounting magnets
  470329-024
 :  305202
Omnimed® Mounting Magnets
  470329-024
Omnimed® Mounting Magnets
Omnimed® Mounting Magnets
Mounting magnets
$47.49
Pack of 4
No more drilling!
  • 1.25" diameter magnets
  • Fits items with a mounting depth of up to 0.10"
Omnimed® mounting magnets are perfect for attaching glove boxes, wall pockets, or any item with standard screw mounting holes to a metal surface. Each magnet is rated to 16 lbs. of force to ensure a secure hold. Each magnet contains a threaded hole and a truss head screw. Simply put the screw through the mounting hole and tighten securely.
 : Pack of 4.
  • Description:
    Mounting magnets