Tube Connector for Combisart® Manifolds, Sartorius
76409-176
- Description:Tube connector
- Cat. No.:76409-176
- Fits:½ L suction flask
This connector is used for connecting a Combisart® stainless-steel manifold to a suction flask 1 or 2 L.
This connector is used for connecting a Combisart® stainless-steel manifold to a suction flask 1 or 2 L.