Diversey™ Oxivir® TB Disinfectant Solution
Supplier: Antyila Scientific
Certificates
About this item
Deliver fast, effective disinfectionin a simple, ready to use liquid
- 1-step cleaner disinfectant powered by AHP technology
- Effective against key pathogens TB, MRSA, norovirus and VRE
- Kills HBV, HCV and HIV in 1-minute, tuberculocidal in 5-minutes and fungicidal in 10-minute
- No VOCs, no added fragrance and no NPEs, environmentally responsible active ingredient breaks down to oxygen and water
Ready-to-use disinfectant cleaner is based on proprietary hydrogen peroxide (AHP) technology to deliver fast, effective cleaning performance. Effective against virucidal, bactericidal, fungicidal, tuberculocidal, and 27 micro-organisms including MRSA and VRE. Kills viruses in 1 minute and sanitises soft surfaces such as carpets, cushions and curtains in 30 seconds. Colorless with a characteristic scent.
Frequently Bought Together