Bond Elut™ Certify delivers a cation exchange/mixed-mode SPE approach that is ideal for extraction of drugs from urine and blood, along with many other applications.
- Special mixed-mode sorbent bed
- Broad application range for aqueous extraction
- Bimodal, nonpolar, and strong cation exchange
The Bond Elut™ Certify extraction cartridge utilizes a packed bed consisting of a special, nonpolar C8 sorbent and a strong cation exchanger (SCX). Certify is most commonly used to extract basic (cationic) drugs from urine and blood, but it is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.
Request the Agilent Bond Elut™ Certify and Ceritfy II Methods Manual here. For forensic use.