ContactUsLinkComponent
3D Culture HTS Cell Viability Assay Kit (Colorimetric), BioVision
3D Culture HTS Cell Viability Assay Kit (Colorimetric), BioVision
Catalog # 76415-410
Supplier:  Avantor
CAS Number:  
3D Culture HTS Cell Viability Assay Kit (Colorimetric), BioVision
Catalog # 76415-410
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    3D Culture HTS Cell Viability Assay Kit (Colorimetric)
  • Cat. No.:
    76415-410
  • No. of assays/kit:
    100 assays
Specifications
About this item
For sensitive quantification of viable cells.
  • Non-radioactive
  • High-throughput method for characterizing and screening cell viability and cytotoxicity
  • Easy-to-use
Three-dimensional (3D) cell cultures are artificially-created environments, where cells are allowed to grow or interact with their surroundings in a 3D fashion. 3D cell cultures improve the function, differentiation and viability of cells and recapitulate the in vivo microenvironment as compared to conventional 2D cell cultures. 3D matrices provide a physiologically relevant screening platform by mimicking the in vivo responses for many cell types including cancer and stem cells. This is specifically important in developmental morphogenesis, pharmacology, drug metabolism and drug toxicity studies. Additionally, quantification of the number of viable cells is an indispensable tool in in vitro screening in these studies. However, with the use of 3D matrices, some protease-based dissociation methods don’t completely dissolve the matrices or the cell aggregates, which may alter the result of the in vitro viability assessment quantitatively.
BioVision’s 3D Culture Cell Viability Assay Kit provides a standardized colorimetric method for sensitive quantification of viable cells that can detect as high as 250,000 viable cells and as low as 1000 viable cells in each well. The absorbance is measured at 460 nm. The measured intensity is proportional to the number of viable cells. Further, the kit comes with an optimized and gentle non-enzymatic dissociation solution for the recovery of viable and dead cells from spheroids in matrices and scaffolds. This assay kit provides an easy-to-use, non-radioactive, high-throughput method for characterizing and screening cell viability and cytotoxicity.