D-Sub Conductive Connector Cover, Menda
Catalog # 76407-246
Supplier: Desco
Specifications
- Description:D-Sub conductive connector cover, 37P (Shell size C/4)
- Cat. No.:76407-246
Specifications
About this item
A cap to cover electrictical connectors.
- Helps elimate dust and other particles in the connector
- Made of conductive material
- Ideal for clean applications
- Free of carbon particulates; will not leave black marks
- Noncorrosive
- Contacts: 37
- Connector type: Pin
Permanently volume conductive and free of carbon black particulate. The connector covers do not leave black marks and are more suited to clean situations. Meets the requirements of NASM5501 31/32. Connector covers are noncorrosive and produce no visible corrosion when tested to Federal Test Method 101C using Kovar test plates. The material used in the covers was selected for its resilience. Properly sized, the covers provide a snug, secure fit. This helps keep connectors clean and prevents the cover from accidentally falling off.