About this item
VWR® Real Time Electrophoresis Systems have revolutionized DNA electrophoresis. It is no longer necessary to remove a gel from a tank, place it onto a transilluminator for viewing or place the gel into an imaging system to take a photo. These versatile and compact systems are an all-in-one replacement for a power supply, gel tank, transilluminator and gel imaging system.
- Monitor DNA separation in real time
- Included blue LED transilluminator slides under the gel tank
- Amber emission filter is built into the gel tank safety lid
- Imaging enclosure included for smart phone documentation
- Includes 2 casting gel and combs
- Choice of 2 versions: With or without power supply
The small but powerful blue LED transilluminator optimally excites safe green DNA stains such as SYBR® Green, Gel Green™ and others. The 465 nm blue wavelength is safer than UV and will not damage DNA in the gels. The amber emission filter in the gel tank cover contacts the running buffer to prevent fogging so that DNA bands can be clearly visualized as they separate. The smart phone imaging enclosure, fits on top of the tank lid and allows quick imaging of separation results using a smart phone. All iPhone, Samsung and other popular phones are supported.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76365-596
- 76359-756
- 76359-758
- Model:
- Without Power Supply
- With Power Supply
- With Power Supply
- Buffer Volume:
- 200 - 225 ml
- 200 - 225 ml
- 200 - 225 ml
- UV Wavelength:
- 465 nm
- 465 nm
- 465 nm
- Tank W×D×H:
- 120×110×45 mm (inner dimensions)
- 120×110×45 mm (inner dimensions)
- 120×110×45 mm (inner dimensions)
- Unit Dimensions:
- 25.5×21×14 cm (10×8.3×5")
- 25.5×21×14 cm (10×8.3×5")
- 25.5×21×14 cm (10×8.3×5")
- Voltage:
- 35, 50 or 100 V
- 35, 50 or 100 V
- Weight:
- 1.04 kg (2.3 lbs.)
- 1.04 kg (2.3 lbs)
- 1.04 kg (2.3 lbs)