Order Entry
United States
ContactUsLinkComponent
Bertin Instruments Anti-rotation Kit for Precellys 24, Bertin Corp
Bertin Instruments Anti-rotation Kit for Precellys 24, Bertin Corp
Catalog # 76415-490
Supplier:  BERTIN CORP
Bertin Instruments Anti-rotation Kit for Precellys 24, Bertin Corp
Catalog # 76415-490
Supplier:  BERTIN CORP
Supplier Number:  S000704-PR240-A.0

Specifications

  • Description:
    Antirotation kit
  • Cat. No.:
    76415-490
  • For use with:
    Precellys 24 homogeniser

Specifications

About this item

The Precellys 24 anti-rotation kit allows you to replace the anti-rotation system in your Precellys 24.

  • Contains all the pieces to replace your anti-rotation system

Designed to enhance the lifetime of your Precellys 24 equipment.