Order Entry
United States
ContactUsLinkComponent
Serum-Free Recombinant RocketFuel™, Cell Systems
Serum-Free Recombinant RocketFuel™, Cell Systems
Catalog # 103810-718
Supplier:  Cell Systems
CAS Number:  
undefined
Serum-Free Recombinant RocketFuel™, Cell Systems
Catalog # 103810-718
Supplier:  Cell Systems
Supplier Number:  SF4ZR500R
CAS Number:  

Specifications

  • Description:
    Reagent
  • Size:
    8 ml
  • Cat. No.:
    103810-718

Specifications

About this item

Obtain better results using Cell Systems growth supplements when initiating, expanding or passaging cultures.

  • High quality specialized reagents
  • Always available
  • Overnight delivery

RocketFuel-R™ supplements 500mL Cell Systems SF-4Z0-500-R. RocketFuel-R™ is the broad-spectrum mitogen growth supplement containing recombinant human growth factors and has been designed to provide defined growth supplements in the serum-free media environment.

Handling and Storage: This product is delivered at ambient temperature. Use 8ml to supplement 500 ml cell culture medium. If the entire unit will not be used within 30 days, aliquot and freeze. Once opened, shelf life is 30 days at 2 to 8 °C.

Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.