Order Entry
United States
ContactUsLinkComponent
Serum-Free RocketFuel™, 8 ml, Cell Systems
Serum-Free RocketFuel™, 8 ml, Cell Systems
Catalog # 103810-716
Supplier:  Cell Systems
CAS Number:  
undefined
Serum-Free RocketFuel™, 8 ml, Cell Systems
Catalog # 103810-716
Supplier:  Cell Systems
Supplier Number:  SF4ZR500
CAS Number:  

Specifications

  • Description:
    Reagent
  • Size:
    8 ml
  • Cat. No.:
    103810-716

Specifications

About this item

Obtain better results using Cell Systems growth supplements when initiating, expanding or passaging cultures.

  • High quality specialized reagents
  • Always available
  • Overnight delivery

RocketFuel™ Supplement. The 8 ml vial activates 500 ml Cell Systems Serum-Free Medium.

RocketFuel™ is the broad-spectrum mitogen growth supplement designed for use with Serum-Free Medium. RocketFuel™ contains Cell Systems bovine growth factor and has been designed to supplement growth in the serum-free media environment. RocketFuel™ is included in various Cell Systems Medium Kits (please see Serum-Free Medium Kits for more information), and is also available individually as a stand-alone supplement to improve performance of your serum-free medium.

Handling and Storage: This product is delivered at ambient temperature. Use 8ml to supplement 500 ml cell culture medium. If the entire unit will not be used within 30 days, aliquot and freeze. Once opened, shelf life is 30 days at 2 to 8 °C.

Certifications: Cell Systems media and reagents are Sterile, made with WFI and all components are cGMP and ISO Compliant.