Order Entry
United States
ContactUsLinkComponent
Cell Systems Human Glomerular Microvascular Endothelial Cells, Primary Cell and Media Company
Cell Systems Human Glomerular Microvascular Endothelial Cells, Primary Cell and Media Company
Catalog # 103810-638
Supplier:  Cell Systems
CAS Number:  
undefined
Cell Systems Human Glomerular Microvascular Endothelial Cells, Primary Cell and Media Company
Catalog # 103810-638
Supplier:  Cell Systems
Supplier Number:  ACBRI128
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Single Donor/Pooled:
    Single donor
  • Description:
    Human glomerular microvascular endothelial cells
  • Size:
    Vial
  • Storage Conditions:
    Upon receipt, store in Liquid Nitrogen
  • Shipping Conditions:
    Shipped on Dry Ice
  • Storage Buffer:
    Frozen in Cell Systems Cell Freezing Medium (4Z0705)
  • Cat. No.:
    103810-638

Specifications

About this item

Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.

  • Highly proliferative and healthy (~85% viability)
  • Antibody free and more biologically relevant
  • Offered with high cell count (~1 million cells per vial)
  • Well-cited in scientific journals
  • Ideal for 2D and 3D cultures, microfluidic devices and co-cultures

Primary Human Glomerular Microvascular Endothelial Cells (ACBRI128) were initiated by decapsulated decapsulated glomeruli isolated from normal human kidney cortical tissue. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.

These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (< 12 cumulative population doubling) cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.

Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.

Standard Tests:
HIV Serologic Test (donor level HIV AB EIA) -Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Hepatitis B (HBV) PCR Test (frozen cells) - Negative
Hepatitis C (HCV) PCR Test (frozen cells) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.

Characterization:
Inducible expression of CD 62E (E-Selectin) < 2% positive by immunofluorescence
Cytoplasmic Vimentin Intermediate Filaments > 98% positive by immunofluorescence
Cytoplasmic Alpha-Actinin Microfilaments > 98% positive by immunofluorescence
Cytoplasmic Fibronectin  > 98% positive by immunofluorescence.

Caution: For Research Use Only