ContactUsLinkComponent
Blue Phantom® Transesophageal and Transthoracic Echo Ultrasound Trainer
Blue Phantom® Transesophageal and Transthoracic Echo Ultrasound Trainer
Catalog # 470327-442
undefined
Blue Phantom® Transesophageal and Transthoracic Echo Ultrasound Trainer
Catalog # 470327-442
Supplier Number:  BPTEE1702
Specifications
  • Description:
    Echo ultrasound trainer
  • Cat. No.:
    470327-442
Specifications
About this item
High-quality, proven technology.
  • Extremely realistic imaging
  • Feels and cannulates like real human tissue
  • Heart contains accurate anatomy
  • Excellent for 2-D, 3-D, and 4-D training
  • Extremely durable
  • Contains anatomical landmarks
  • Use any ultrasound imaging system
Excellent for training clinicians in the techniques associated with transthoracic echocardiography, transesophageal echo (TEE), and ultrasound guided pericardiocentesis procedural training. Blue Phantom®'s TTE/TEE cardiac ultrasound training platform allows users to develop and practice ultrasound imaging skills as TEE, transthoracic echocardiography, and ultrasound guided pericardiocentesis.
These skills include; using ultrasound system controls, transducer insertion and placement, probe positioning and movement, applying proper transducer pressure to obtain images, recognition of the cardiac structures, lung and rib artifacts and other anatomical structures, and guiding needles and catheters to the pericardial space for pericardiocentesis training. provides an extremely realistic echocardiography training platform as you utilize your own ultrasound system for simulation training. Self-healing simulated tissue allows for repeated procedural training without fluid leakage while utilizing 18 - 21 gauge needles and corresponding catheters.
This extremely realistic life size transesophageal TEE and transthoracic echo simulator includes an articulating jaw and extends from the head to the mid thigh. Utilize your own ultrasound system and a TEE simulator; this simulation trainer will perform well using any ultrasound imaging system configured with appropriate transducers for echocardiography imaging procedures.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.