Prepacked validation columns are suitable for scale-down studies such as viral clearance.
- High-quality packing
- Excellent dimensions for scale-down studies
- Prepacked with the most commonly used BioProcess resins
- High consistency between columns in terms of efficiency and asymmetry
Results of Analysis certificate available for each individual column - includes column performance testing such as asymmetry and efficiency. Developed together with biopharmaceutical experts to ensure optimal function for scale-down studies such as viral clearance.
They feature a high-quality packed bed, enabling robust and reproducible process development or validation. Columns available for custom packing include Tricorn, XK, and HiScale.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 103743-748
- 103743-746
- 103743-750
- 103743-738
- 103743-744
- 103743-736
- 103743-734
- pH Range:
- 2 - 14
- 2 - 14
- 2 - 14
- 2 - 14
- 2 - 14
- 2 - 14
- 2 - 14
- Storage:
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol