Practum® Balances, Precision Models, Sartorius
Supplier: Avantor
Certificates
About this item
A practical basic balance that incorporates Sartorius' renowned weighing technology to deliver reliable professional results. Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting, and averaging. Results can be transferred via a mini-USB interface to a printer or a computer directly into Microsoft Excel® spreadsheet without additional software. Menu with choice of 10 languages.
- High contrast colour touch screen
- Automatic calibration and adjustment possible with an optional external weight
- Robust housing design and rugged weigh cell
- Supervisor lock to prevent any unintentional changes to settings
- Built-in overload protection
Delivery: Supplied with universal power adapter, in-use cover and application guide but without calibration weight.
Frequently Bought Together