About this item
Ultra Uniform Gold Nanospheres for applications requiring the highest uniformity in size and shape
- For applications requiring the highest uniformity in size and shape
Our Ultra Uniform particles have the lowest coefficients of variation available anywhere. They are used as particle size and concentration standards, electron microscopy standards, and for applications where tight tolerances on the particle size, shape, mass, or volume are required. Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch to specific COA's with TEM, DLS, Zeta, and UV to Vis.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.