About this item
Purified, concentrated, high quality gold nps, sterile and endotoxin-free. Near-netural PEG surface
- High quality nanoparticles, precisely engineered and highly characterized
- Unagglomerated and monodisperse
- High concentration materials (1 mg/ml, 5.08 mm, 1000 ppm)
- Extensively purified to remove residual reactants
- Manufactured sterile; provided in ultra to pure (Milli to Q) DI water
- BioPure mateirals ideal for in to vitro and in to vivo experiments, nanotoxicology
- PEG surface provides strong steric stability in salts and alcohols
Spherical gold nanoparticles, 1 mg/ml (high concentration), PEG surface provides strong steric stability in salts and alcohols. BioPure material is Sterile and Endotoxin to Free (<5 EU/ml). Precisely engineered, unagglomerated nanospheres with narrow size distributions (CVs <12% for 7 to 15 nm sizes, <15% for 20 to 100 nm sizes, and <18% for 5 nm size).
Each batch extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products provided with batch-specific COA's with TEM, DLS, Zeta, and UV-Vis measurements.
Colloidal gold has applications in Diagnostics, Lateral Flow Assays, Molecular Detection, Photothermal and Microwave-based Therapies, TEM labels, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), and novel Color Enhancements and Pigments.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.