ContactUsLinkComponent
API 50 CHB/E Medium, bioMérieux
API 50 CHB/E Medium, bioMérieux
Catalog # 97060-444
Supplier:  bioMerieux
CAS Number:  
API 50 CHB/E Medium, bioMérieux
Catalog # 97060-444
Supplier:  bioMerieux
Supplier Number:  50430
CAS Number:  
Specifications
  • Description:
    API® 50 CHB Medium
  • Cat. No.:
    97060-444
  • Volume:
    10× 10 ml
  • Supplier no.:
    50430
Specifications
About this item
API 50 CHB/E Medium is intended for the identification of Bacillus and related genera, as well as Gram-negative rods belonging to the Enterobacteriaceae and Vibrionaceae families. It is a ready-to-use medium which allows the fermentation of the 49 carbohydrates on the API 50 CH strip to be studied.