Order Entry
United States
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB)

3,3',5,5'-Tetramethylbenzidine (TMB)

3,3',5,5'-Tetramethylbenzidine (TMB)
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB)

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

TMB ELISA Peroxidase Substrate (3, 3', 5, 5' - Tetramethylbenzidine) is a chromogenic substrate used to visualize antibody reactivity in ELISA experiments. In the presence of peroxidases, TMB can act as an electron donor for the conversion of peroxides to water, changing the color of solution to a blue color equivalent to the degree of reactivity. This reaction can be halted with acid to change the TMB to yellow. TMB ELISA Peroxidase Substrate is ideal for investigators in Microbiology, Immunology, and Cancer research.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 8 L
    Log in for availability
    Log in for availability
    Each (8l)
    Each (8l) - $1,772.44
    $1,772.44
    Each (8l)
       $1,772.44
    Minimum order quantity is 1

    Supplier Number: TMBE-8000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 20 L
    Log in for availability
    Log in for availability
    Each (20l)
    Each (20l) - $4,441.64
    $4,441.64
    Each (20l)
       $4,441.64
    Minimum order quantity is 1

    Supplier Number: TMBE-20L
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.