About this item
Kits for increasing immunofluorescence sensitivity using HRP-catalyzed tyramide signal amplification with the choice of next-generation fluorescent CF® dyes or biotin.
- Increase sensitivity by up to 100-fold
- Use lower primary antibody concentrations
- Boost signal in challenging samples, like human FFPE tissues
- Choose from six bright and photostable CF® dyes or biotin
Tyramide amplification, sometimes called Catalyzed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to conventional avidin-biotin detection. Multiple rounds of tyramide signal amplification can be performed for multiplex detection.
The HRP goat anti-rabbit antibody provided in the kits has been adsorbed against human, mouse, and rat serum to minimize cross-reactivity. The kits include choice of six CF® dye tyramides or biotin tyramide. CF® dyes are line of next-generation fluorescent dyes, with advantages in brightness and photostability compared to other dyes.