Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:RNAstorm™ RNA isolation kit
- Size:50 preparations
- Description:RNAstorm™ RNA isolation kit
- Cat. No.:76404-074
The RNAstorm™ kit provides a convenient and efficient method of isolating RNA from fresh or frozen cells or tissue samples.
- High quality total RNA in as little as 20 minutes
- Simple workflow yielding up to 120 μg of RNA
- DNase treatment included
- No phenol-chloroform or ethanol precipitation
- Isolate RNA for next-gen sequencing (RNA-Seq), RT-PCR, cDNA synthesis, or microarrays
Purify high quality total RNA from either cultured cells or tissue (fresh or fresh/frozen) in as little as 20 minutes. High yields (up to 120 μg) can be obtained using a simple and quick column-based protocol. Contaminating DNA is removed using a DNase treatment step, and the protocol avoids toxic chemicals such as phenol or chloroform.