Masterflex® L/S® Precision Pump Tubing, Norprene® Food A-60-F, Cole Parmer
: Avantor
This tubing is designed for flexibility at extreme temperatures.
- Heat, ozone, and UV light resistant
- Nonaging, nonoxidizing
- Superior acid and alkali resistance
- Opaque beige
- Temperature range: –51 to 132 °C (−60 to 270 °F)
- Sterilization: Autoclave, ethylene oxide, or gamma irradiation
Applications: Ideal for the food, dairy, and beverage industries.
: FDA, Non DEHP, Non Phthalate, RoHS.
Frequently Bought Together