Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL-33 (from E. coli)
Human Recombinant IL-33 (from E. coli)
Catalog # 103705-358
Supplier:  Novus Biologicals
CAS Number:  
Human Recombinant IL-33 (from E. coli)
Catalog # 103705-358
Supplier:  Novus Biologicals
Supplier Number:  3625-IL-010/CF
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    10 µg
  • Storage Conditions:
    2...8 °C (1 month from the date of receipt);–20...–70 °C (3 - 12 months from the date of receipt)
  • Endotoxin Content:
    <0.01 EU per 1 μg of the protein by the LAL method
  • Biological Activity:
    Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. The ED50 for this effect is 0.06 - 0.24 ng/ml
  • Reconstitution Instructions:
    Reconstitute at 10 μg/ml in sterile PBS
  • Carrier-Free:
    Y
  • Protein Synonyms:
    interleukin 33|Nuclear factor from high endothelial venules|interleukin-33|IL-1F11|DVS27-related protein|IL33|C9orf26|IL1F11|NF-HEV|NF-HEVNFEHEV|DKFZp586H0523|DVS27|C9orf26chromosome 9 open reading frame 26 (NF-HEV)|NFHEV|Interleukin-1 family member 11|RP11-575C20.2
  • Accession No.:
    O95760
  • UniProtKB:
    O95760
  • Protein/Peptide Name:
    IL-33
  • Purity:
    >95%
  • Molecular Weight:
    18 kDa
  • Sequence:
    Ser112
  • Formulation:
    Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT
  • Tested Applications:
    Bioactivity
  • Purification:
    SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining
  • Cat. No.:
    103705-358

Specifications

About this item

IL-33, also known as NF-HEV and DVS 27, is a 30 kDa proinflammatory protein that may also regulate gene transcription (1‑3).

DVS 27 was identified as a gene that is upregulated in vasospastic cerebral arteries. NF‑HEV was described as a nuclear factor that is preferentially expressed in the endothelial cells of high endothelial venules relative to endothelial cells from other tissues. IL‑33 was identified based on sequence and structural homology with IL‑1 family cytokines.

DVS 27, NF‑HEV, and IL‑33 share 100% amino acid sequence identity. IL‑33 is constitutively expressed in smooth muscle and airway epithelia. It is up-regulated in arterial smooth muscle, dermal fibroblasts, and keratinocytes following IL‑1 alpha or IL‑1 beta stimulation (1, 3). Similar to IL‑1, IL‑33 can be cleaved in vitro by caspase‑1, generating an N‑terminal fragment that is slightly shorter than the C‑terminal fragment (3, 4).

The N‑terminal portion of full length IL‑33 contains a predicted bipartite nuclear localization sequence and a homeodomain‑like helix‑turn‑helix DNA binding domain. By immunofluorescence, full length IL‑33 localizes to the nucleus in HUVECs and transfectants. The C‑terminal fragment, corresponding to mature IL‑33, binds and triggers signaling through mast cell IL‑1 R4/ST2L, a longtime orphan receptor involved in the augmentation of Th2 cell responses (3, 5‑7).

A ternary signaling complex is formed by the subsequent association of IL‑33 and ST2L with IL‑1R AcP. Stimulation of Th2 polarized lymphocytes with mature IL‑33 in vitro induces IL‑5 and IL‑13 secretion. In vivo administration of mature IL‑33 promotes increased production of IL‑5, IL‑13, IgE, and IgA, as well as splenomegaly and inflammatory infiltration of mucosal tissues. Full length and mature human IL‑33 share 52 ‑ 58% aa sequence identity with mouse and rat IL‑33. Human IL‑33 shares less than 20% aa sequence identity with other IL‑1 family proteins.