ContactUsLinkComponent
Tek-Select® Bouin’s Fixative, IMEB
Tek-Select® Bouin’s Fixative, IMEB
Catalog # 103783-074
Supplier:  IMEB INC MS
CAS Number:  
undefined
Tek-Select® Bouin’s Fixative, IMEB
Catalog # 103783-074
Supplier:  IMEB INC MS
Supplier Number:  S129
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Bouin's fixative
  • Size:
    1 Gallon
  • Cat. No.:
    103783-074
Specifications
About this item
Tek-Select® Bouin’s Fixative is ideally suited for gastrointestinal tract biopsies because it allows crisper and better nuclear staining than 10% neutral-buffered formalin.
  • Crisper and better nuclear staining
  • Packaged in one gallon bottle
This fixative is also excellent when tissue structure with a soft and delicate texture must be preserved.