Order Entry
United States
ContactUsLinkComponent
Tek-Select® Cassette Filing Cabinet, 6 Drawers, IMEB
Tek-Select® Cassette Filing Cabinet, 6 Drawers, IMEB
Catalog # 103783-072
Supplier:  IMEB INC MS
undefined
Tek-Select® Cassette Filing Cabinet, 6 Drawers, IMEB
Catalog # 103783-072
Supplier:  IMEB INC MS
Supplier Number:  4192TS

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Width:
    9½"
  • Capacity:
    1500 cassettes or 1000 rings
  • Description:
    Cassette filing cabinet
  • Height:
    17"
  • Length:
    15"
  • Cat. No.:
    103783-072

Specifications

About this item

The Tek-Select® Cassette Filing Cabinet provides a permanent storage reference unit for cassettes and rings. The filing cabinets are stackable and accommodate 1,500 cassettes or 1000 rings.

  • Stackable design
  • Six drawers
  • Hold 1500 cassettes or 1000 rings
  • One-piece construction

Tek-Select® Cassette Filing Cabinets feature one-piece construction to eliminates hinges and doors for dust-free storage. The cabinet includes 6 impact-resistant plastic drawers with identification area writing surfaces.