Specifications
- Description:Derm freezing medium
- Size:12×4 oz.
- Cat. No.:103783-070
Specifications
About this item
Specifically formulated for pathology cryostat tissue sectioning, IMEB’S DFM (Derm Freezing Medium) formulation freezes at optimum cutting temperature, creating a secure hold of all tissue types.
- Freezes at optimum cutting temperature
- Fast binding of all fresh tissue
- Flatter sectioning
- Low viscosity
- Clear
Use of IMEB’S DFM (Derm Freezing Medium) formulation ensures fast binding of all fresh tissue. This unique formula also reduces 'medium roll' for flatter sectioning.