Order Entry
United States
ContactUsLinkComponent
Tek-Select® Diff-Stain Kit, IMEB
Tek-Select® Diff-Stain Kit, IMEB
Catalog # 103783-058
Supplier:  IMEB INC MS
CAS Number:  
undefined
Tek-Select® Diff-Stain Kit, IMEB
Catalog # 103783-058
Supplier:  IMEB INC MS
Supplier Number:  K7128
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Diff-stain kit
  • Cat. No.:
    103783-058

Specifications

About this item

The Tek-Select® Diff-Stain kit is designed to provide Nuclear and cytoplasmic staining and recently has provided excellent staining for Helicobacter pylori as well as a Hematology stain.

  • 8 oz. bottle of Solution A (fixative)
  • 8 oz. bottle of Solution B (azure stain)
  • 8 oz. bottle of Solution C (xanthene dyes)
  • Three 2 oz. staining containers
  • Storage tray

The Diff-Stain kit results in Nuclear staining from blue to purple, and pink to orange with paraffin and frozen section staining. Helicobacter pylori provides a deep rich purple. This stain is also excellent as a hematology stain.

Ordering information: Kit includes 8 oz. bottles of solution A (fixative), solution B (azure Stain), solution C (xanthene dyes), three 2 oz. staining containers, and a storage tray.