Order Entry
United States
ContactUsLinkComponent
Tek-Select® Tissue Marking Dyes Multi Mix Starter Kit, IMEB
Tek-Select® Tissue Marking Dyes Multi Mix Starter Kit, IMEB
Catalog # 103783-054
Supplier:  IMEB INC MS
CAS Number:  
undefined
Tek-Select® Tissue Marking Dyes Multi Mix Starter Kit, IMEB
Catalog # 103783-054
Supplier:  IMEB INC MS
Supplier Number:  TSMP-6
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Tissue marking dyes multi mix starter kit
  • Size:
    (6) 2 oz. Bottles
  • Cat. No.:
    103783-054

Specifications

About this item

Tek-Select® tissue marking dyes are excellent for marking excised tissue margins or for processing multiple specimens in one tissue cassette.

  • Six colors packaged in 2 oz. bottles
  • Application sticks included

Tek-Select® tissue marking dyes are excellent for marking excised tissue margins or for processing multiple specimens in one tissue cassette. Tissue marking dyes are fast drying with excellent readability, and are supplied ready to use.