About this item
Versatile, durable corrugated cardboard boxes are strong yet lightweight, with a moisture-resistant exterior wax coating.
- Two sets of five drawers
- Total capacity of 5000 cassettes per case of 10 drawers
- Strong yet lightweight construction
- Moisture-resistant with a special wax coating
- Outer corrugated shell acts as a stabilizer for stacking
- Available pre-assembled or unassembled
Tek-Select® Cardboard Cassette Storage Files offer a less expensive option to metal and plastic storage filing systems. Boxes can be purchased pre-assembled or unassembled. An outer corrugated shell acts as a stabilizer in keeping units stacked together over a long period of time. Each box section accommodates 500 cassettes. With 10 boxes per case the total capacity of each case is 5000 cassettes.