ContactUsLinkComponent
Polypropylene Rinse Container, PerkinElmer
Polypropylene Rinse Container, PerkinElmer
Catalog # 10139-284
Supplier:  PerkinElmer
undefined
Polypropylene Rinse Container, PerkinElmer
Catalog # 10139-284
Supplier:  PerkinElmer
Supplier Number:  N0777259
Specifications
  • Capacity:
    4 L
  • Description:
    Rinse container
  • Material:
    PP
  • Cat. No.:
    10139-284
  • Supplier no.:
    N0777259
Specifications
About this item
This rinse container, labeled rinse 2, does not contain a filtered inlet. A rinse container with filtered inlet is available separately.
Delivery: Cap for rinse container is included.