ContactUsLinkComponent
Quartz Torch Bonnet for Optima™ 3×00 XL/3×00 DV, PerkinElmer
Quartz Torch Bonnet for Optima™ 3×00 XL/3×00 DV, PerkinElmer
Catalog # 97012-910
Supplier:  PerkinElmer
undefined
Quartz Torch Bonnet for Optima™ 3×00 XL/3×00 DV, PerkinElmer
Catalog # 97012-910
Supplier:  PerkinElmer
Supplier Number:  N0691664
Specifications
  • Description:
    Quartz torch bonnet for Optima™ 3×00 XL/3×00 DV
  • Cat. No.:
    97012-910
  • Supplier no.:
    N0691664
Specifications
About this item
This quartz bonnet is used with Optima™ 3×00 XL and 3×00 DV ICP-OES series instruments.
The torch bonnet is used as a spacer between the torch and RF load coil assembly.