F5-50 Solution Fitting for Standard MEINHARD™ Nebulizers, PerkinElmer
10139-384
- Description:F5-50 Solution fitting for MEINHARD Nebs
- Cat. No.:10139-384
- Supplier no.:N0777414
This Standard Nebulizer Solution Fitting is designed for use with Optima™ 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments.
- PTFE capillary measures 0.5 mm I.D.×70 cm in length
This nebulizer fitting is included with all standard MEINHARD™ Glass Nebulizers.