Sample Capillary Adapter for GemCone and GemTip Nebulizers, PerkinElmer
Catalog # 97012-744
Supplier: PerkinElmer
Specifications
- Description:Sample capillary adapter for nebulizers
- Cat. No.:97012-744
- Supplier no.:N0371505
Specifications
About this item
This Capillary Adapter is designed for use with AAnalyst® 100/300/200/400/700/800 AA series instruments, as well as all Avio and Optima™ ICP-OES series instruments.
This Capillary Adapter, or Gland Nut, is included in the GemTip High-Sensitivity Plastic Body Nebulizer Complete Assembly for AAnalyst® and the Universal GemTip Corrosion-Resistant Plastic Body Nebulizer Complete Assembly for AAnalyst®. It is used in the GemCone Low Flow Nebulizer and the GemCone High Dissolved Solids Nebulizer.