Universal GemTip Capillary Assembly, PerkinElmer
10138-026
- Description:Universal GemTip capillary assembly
- Cat. No.:10138-026
- Supplier no.:N0371520
This Universal GemTip Capillary Assembly is for use with AAnalyst® 100/300/200/400/700/800 and earlier AA series instruments.
- Only includes capillary needle assembly, not complete assembly
This capillary assembly is included in the Universal GemTip Corrosion-Resistant Plastic Body Nebulizer Complete Assembly. It is compatible with Models 5100PC/5000/3300/3110/3100 AA instruments.