ContactUsLinkComponent
Non-Coded Intensitron Lamp Plug Adapter, PerkinElmer
Non-Coded Intensitron Lamp Plug Adapter, PerkinElmer
Catalog # 10138-374
Supplier:  PerkinElmer
undefined
Non-Coded Intensitron Lamp Plug Adapter, PerkinElmer
Catalog # 10138-374
Supplier:  PerkinElmer
Supplier Number:  N3050197
Specifications
  • Description:
    Non-Coded intensitron lamp adapter
  • Cat. No.:
    10138-374
  • Supplier no.:
    N3050197
Specifications
About this item
Adapter allows non-coded Intensitron lamps (03036XXX series, 9-pin) and other 9-pin lamps to be used with PinAAcle and AAnalyst instruments (4-pin).
For use with legacy PerkinElmer Atomic Absorption (AA) instruments with Intensitron lamp connectors.