ContactUsLinkComponent
HGA L`vov Graphite Platforms, PerkinElmer
HGA L`vov Graphite Platforms, PerkinElmer
Catalog # 97012-826
Supplier:  PerkinElmer
undefined
HGA L`vov Graphite Platforms, PerkinElmer
Catalog # 97012-826
Supplier:  PerkinElmer
Supplier Number:  B3001256
Specifications
  • Description:
    HGA L`vov graphite platforms
  • Cat. No.:
    97012-826
  • Supplier no.:
    B3001256
Specifications
About this item
The L'vov platform is a small plate of solid pyrolytic graphite that's inserted into compatible graphite tubes. It has a slight depression in the center which can accommodate up to 50 ml of solution.
  • Vaporization into a higher temperature gas atmosphere producing more free atoms, which reduce interferences
  • Longer tube life through reduced attack by sample matrix and reagents
  • Platforms are compatible with PerkinElmer HGA graphite tubes
The function of the L'vov platform is to isolate the sample from the tube walls to allow more reproducible atomization of the sample through indirect heating. The platform heats primarily by the radiation given off from the tube walls. Sample vaporization and atomization occur after the tube reaches a steady-state temperature.