ContactUsLinkComponent
Demountable Quartz Injector, 1.0 mm Int.Ø., PerkinElmer
  10139-376
undefined
Demountable Quartz Injector, 1.0 mm Int.Ø., PerkinElmer
  10139-376
 :  N0777410
  • Ø int.:
    1.0 mm
  • Description:
    Demountable quartz injector
  • Cat. No.:
    10139-376
  • Supplier no.:
    N0777410
This Quartz Injector Cassette Mount is designed for use with demountable torches in NexION® 300/350 ICP-MS series instruments.
  • Demountable design allows the injector to be removed and replaced efficiently without dismounting the torch
Standard Injectors have an O-Ring free direct connection to Cyclonic and PFA spray chambers while O-Ring free Injectors have an O-Ring free seal that reduces contamination, makes it easy to clean and easy to insert. This injector can be used with the 0-Slot Demountable Quartz Torch (N8122006). A 1.0 mm I.D. O-Ring Free Injector is available separately (101686-298).