ContactUsLinkComponent
Nebulizer O-Ring Kit for 5100, PerkinElmer
Nebulizer O-Ring Kit for 5100, PerkinElmer
  10138-768
undefined
Nebulizer O-Ring Kit for 5100, PerkinElmer
  10138-768
 :  N9300065
  • Description:
    Nebulizer O-ring kit
  • Cat. No.:
    10138-768
  • For use with:
    AAnalyst 100/200/300/400/700/800
  • Supplier no.:
    N9300065
This metal body O-Ring Kit is used in the Stainless Steel, Corrosion-Resistant, and Platinum Alloy nebulizers for AAnalyst® 100/200/300/400/700/800 and earlier AA series instruments.
  • Compatible with the Corrosion-Resistant Nebulizer Complete Assembly for AAnalyst®
  • Compatible with the Platinum Alloy Nebulizer Complete Assembly for AAnalyst®
  • Compatible with the Stainless Steel Nebulizer Complete Assembly for AAnalyst®
  • Compatible with Models 3100/3110/3300/5000/5100 PC AA instruments as wel
 : This kit includes: (5) O-Rings for Insert/Venturi, (3) O-Rings for Capillary Assembly, (2) O-Rings for front end cap, and (2) Solvent Resistant O-Rings for front end cap.