Order Entry
United States
ContactUsLinkComponent
Carbon Fiber Sample Probe, 10" for ASXPRESS® PLUS, PerkinElmer
Carbon Fiber Sample Probe, 10" for ASXPRESS® PLUS, PerkinElmer
  10139-504
undefined
Carbon Fiber Sample Probe, 10" for ASXPRESS® PLUS, PerkinElmer
  10139-504
 :  N0777568

 

  • Ø int.:
    1.0 mm
  • Description:
    10" Carbon fiber sample probe
  • Cat. No.:
    10139-504
  • For use with:
    ASXPRESS® PLUS
  • Supplier no.:
    N0777568

 

 

Carbon fiber sample probe, with Int.Ø of 1.0 mm, for the ASXPRESS® PLUS.

The ASXPRESS® PLUS FEP-covered carbon fiber sample probe is 10" in length, and with tubing, has a capillary length of 75".

 : This probe kit includes PFA capillary tubing, capillary tube connection fitting, (2) probe clamps, ferrule, and (5) O-Rings.