PolyPro-ST Nebulizer for Optima™ 2x00/4x00/5x00/7x00/8x00 Instruments, PerkinElmer
Catalog # 10138-238
Supplier: PerkinElmer
Specifications
- Description:PolyPro-ST nebulizer for Optima™ instruments
- Cat. No.:10138-238
- Supplier no.:N0777219
Specifications
About this item
The PolyPro-ST nebulizer with detachable capillary is designed for use with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV, and 8x00 ICP-OES series instruments.
- Polypropylene construction
- Hydrofluoric acid resistant
- 0 to 3 ml/min. sample flow rate with peristaltic pumping
- Resistant to clogging and accidental breakage
The PolyPro-ST Nebulizer is an HF resistant alternative to the PFA nebulizer, but has the same high efficiency aerosol generation. It is a robust nebulizer that can self-aspirate with an exchangeable external sample uptake capillary or be used with a peristaltic pump.