Radial Purge Extension Window for Optima ICP-OES Instruments, PerkinElmer
Catalog # 97013-462
Supplier: PerkinElmer
Specifications
- Description:Radial purge extension window for optima
- Cat. No.:97013-462
- Supplier no.:N0581497
Specifications
About this item
This quartz radial window for the purge extension is used with Optima™ 3000, 3000 SCR, 3x00 RL, 4300 V, 5300 V, and 7300 V ICP-OES series instruments.
The radial view purge extension window tube is a replaceable quartz tube that slides into the mount between the torch and the optics for the radial view.