ContactUsLinkComponent
Lanthanum (La) Lumina™ Hollow Cathode Lamp, PerkinElmer
Lanthanum (La) Lumina™ Hollow Cathode Lamp, PerkinElmer
Catalog # 45603-306
Supplier:  PerkinElmer
undefined
Lanthanum (La) Lumina™ Hollow Cathode Lamp, PerkinElmer
Catalog # 45603-306
Supplier:  PerkinElmer
Supplier Number:  N3050141
Specifications
  • Description:
    Lanthanum (La) lumina HCL
  • Cat. No.:
    45603-306
  • Supplier no.:
    N3050141
Specifications
About this item
Single Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Lanthanum (La). Series N30501XX Lumina™ 2" (50 mm) diameter single-element lamps are designed to be used with our PinAAcle™ and AAnalyst™ Atomic Absorption spectrometer series of instruments.
Please note that this element requires the use of a nitrous oxide-acetylene flame. Every genuine , PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime.
Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.