Sample Loops, connect to ports #1 and #4, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
High volume, fast-loading PFA FAST loops for high-flow valves.
Recommended for use with 1.0 mm IØ. (gray) probes and applications requiring larger sample volumes or high flow rates. 3.2 mm OØ. 1/4-28 fittings.
Frequently Bought Together