ContactUsLinkComponent
MicroMist™ Nebulizer for ELAN/NexION 300/350, PerkinElmer
MicroMist™ Nebulizer for ELAN/NexION 300/350, PerkinElmer
  10140-320
undefined
MicroMist™ Nebulizer for ELAN/NexION 300/350, PerkinElmer
  10140-320
 :  N0775341
  • Description:
    MicroMist™ Nebulizer for ELAN/NexION 300/350, uptake rate 0.4 ml/min
  • Cat. No.:
    10140-320
  • Supplier no.:
    N0775341
This glass MicroMist™ nebulizer includes connectors and sample tubing for use with ELAN® 5000/6x00/9000/DRCs and NexION® 300 and 350 ICP-MS series instruments. It can also be used with Optima® 3x00 RL ICP-OES series instruments.
The MicroMist™ micro-uptake glass concentric nebulizer offers high performance for limited sample volumes. MicroMist™ for ELAN/NexION 300/350 are fitted with a UniFit connector with 1.3×0.50×700 mm (I.Ø×O.Ø×L) sample tubing.