Polypropylene Rinse Container (4 L) with Filtered Inlet, PerkinElmer
Catalog # 10139-282
Supplier: PerkinElmer
Specifications
- Capacity:4 L
- Description:Polypropylene rinse container with filtered inlet
- Cat. No.:10139-282
- Supplier no.:N0777258
Specifications
About this item
This 4-liter polypropylene rinse container, labeled Rinse 1, features a filtered air inlet.
Ordering information: The Polypropylene Rinse Container without Filtered Inlet is available separately. Cap for rinse container is included.