Masterflex® I/P® Spooled Pump Tubing, Platinum-Cured Silicone, Cole Parmer
Supplier: Cole Parmer
Certificates
About this item
Excellent biocompatibility that meets multiple classifications. Genuine Masterflex® tubing undergoes robust testing to provide optimal performance and pump life hours with peristaltic pump equipment.
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Excellent heat resistance
- Low compression set, odorless, nontoxic, translucent
- Slightly greater clarity than peroxide-cured tubing
- Smooth surfaces; lower protein binding levels
- Fewer potential leachables
- Sterilize: Autoclave, gamma irradiation
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications.
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Large range of formulations for most extensive chemical compatibility. Single-use custom lengths and configurations available to minimize downtime. Regulatory approvals to match your unique application.
Frequently Bought Together