Masterflex® Transfer Tubings, PTFE, Cole Parmer
: Avantor
The translucent PTFE transfer tubing won't crack under heat or pressure. It is ideal for general-purpose, single-use, and extended fluid path applications.
- Withstands extreme temperatures
- Chemically inert, nontoxic, and virtually nonporous
- Excellent crack- and stress-resistance
- Low coefficient of friction
- Temperature range: –240 to 260 °C (–400 to 500 °F)
- Sterilize: Autoclave, ethylene oxide, or dry heat
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work.
Large range of formulations for most extensive chemical compatibility. Single-use custom lengths and configurations available to minimize downtime. Regulatory approvals to match your unique application.
: Resin is FDA-compliant (21 CFR 177.1550).
Frequently Bought Together